7-Anilino-4-hydroxy-2-naphthalenesulfonic acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Phenyl J acid7-Anilino-4-naphthol-2-sulphonic acid7-Anilino-4-naphthol-2-sulfonic acid

A0467-500G A0467-25G
TCA0467-500GEA 435.69 USD
TCA0467-500G TCA0467-025G
7-Anilino-4-hydroxy-2-naphthalenesulfonic acid ≥95.0% (by titrimetric analysis)
7-Anilino-4-hydroxy-2-naphthalenesulfonic acid

CAS Number: 119-40-4
MDL Number: MFCD00035714
Molecular Formula: C16H13NO4S
Molecular Weight: 315.34
Purity/Analysis Method: 95.0% (T)
Form: Crystal


Other Product Names / Synonyms: 7-Anilino-4-naphthol-2-sulfonic Acid, Phenyl J Acid

Formula: C₁₆H₁₃NO₄S
MW: 315.34 g/mol
MDL Number: MFCD00035714
CAS Number: 119-40-4
https://us.vwr.com/stibo/web/std.lang.all/79/01/28927901.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR