1,2-Bis(2-iodoethoxy)ethane ≥96.0% stabilized
Supplier: TCI America
Synonyms:
Ethylene Glycol Bis(2-iodoethyl) Ether
B1687-10G
TCB1687-010GEA
74.55
USD
TCB1687-010G
1,2-Bis(2-iodoethoxy)ethane ≥96.0% stabilized
1,2-Bis(2-iodoethoxy)ethane
(stabilized with Copper chip)
CAS Number: 36839-55-1
MDL Number: MFCD00075008
Molecular Formula: C6H12I2O2
Molecular Weight: 369.97
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 2.05
Other Product Names / Synonyms: Ethylene Glycol Bis(2-iodoethyl) Ether
Formula:
C₆H₁₂I₂O₂ MW: 369.97 g/mol Density: 2.05 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00075008 CAS Number: 36839-55-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...