Pentyl nitrate ≥98.0%

Supplier: TCI America

Synonyms: Amyl nitratePentyl nitrate

A0453-25ML
TCA0453-025MLEA 273.93 USD
TCA0453-025ML
Pentyl nitrate ≥98.0%
Pentyl nitrate

CAS Number: 1002-16-0
MDL Number: MFCD00059177
Molecular Formula: C5H11NO3
Molecular Weight: 133.15
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 157
Flash Point (°C): 52
Specific Gravity (20/20): 1.00


Product Categories: Bioscience, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: Pentyl Nitrate

Formula: C₅H₁₁NO₃
MW: 133.15 g/mol
Boiling Pt: 157 °C
Melting Pt: –123 °C
Density: 1.00 (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00059177
CAS Number: 1002-16-0
EINECS: 213-684-2
UN: 1112
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/78/91/28927891.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR