(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Boc-Ser(Bzl)-OHN-Boc-O-benzyl-L-serine

B1629-5G
TCB1629-005GEA 40.6 USD
TCB1629-005G
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥98.0% (by HPLC, titration analysis)
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid

CAS Number: 23680-31-1
MDL Number: MFCD00066063
Molecular Formula: C15H21NO5
Molecular Weight: 295.34
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 59
Specific rotation [a]20/D: 22 deg (C=2, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-O-benzyl-L-serine, Boc-Ser(Bzl)-OH

Formula: C15H21NO5
MW: 295.3 g/mol
Melting Pt: 59 °C
MDL Number: MFCD00066063
CAS Number: 23680-31-1
https://us.vwr.com/stibo/web/std.lang.all/33/90/28933390.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR