Bis(4-hydroxy-3,5-dimethylphenyl)sulfone ≥98.0% (by HPLC, titration analysis)

Synonyms: Bis(3,5-dimethyl-4-hydroxyphenyl) Sulfone

B1577-25G
TCB1577-025GEA 44.29 USD
TCB1577-025G
Bis(4-hydroxy-3,5-dimethylphenyl)sulfone ≥98.0% (by HPLC, titration analysis)
Bis(4-hydroxy-3,5-dimethylphenyl)sulfone

CAS Number: 13288-70-5
MDL Number: MFCD00036032
Molecular Formula: C16H18O4S
Molecular Weight: 306.38
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 297


Product Categories: Materials Chemistry, Monomers, Diphenyl Sulfones (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bis(3,5-dimethyl-4-hydroxyphenyl) Sulfone

Formula: C₁₆H₁₈O₄S
MW: 306.38 g/mol
Melting Pt: 297 °C
MDL Number: MFCD00036032
CAS Number: 13288-70-5
https://us.vwr.com/stibo/web/std.lang.all/33/43/28933343.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR