Brilliant black BN

Supplier: TCI America

Synonyms: Black PNBrilliant Black ACI 28440Blue Black BNFood Black 1

B1561-25G
TCB1561-025GEA 154.92 USD
TCB1561-025G
Brilliant black BN
Brilliant black BN

CAS Number: 2519-30-4
MDL Number: MFCD00078980
Molecular Formula: C28H21N5O14S4
Molecular Weight: 867.66
Form: Crystal
Color: Deep Yellow Red
Flash Point (°C): 93
Lambda max.: 572 nm (H2O)


Other Product Names / Synonyms: Brilliant Black PN

Formula: C₂₈H₁₇N₅O₁₄S₄
MW: 867.66 g/mol
Flash Pt: 93 °C
MDL Number: MFCD00078980
CAS Number: 2519-30-4
https://us.vwr.com/stibo/web/std.lang.all/33/29/28933329.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR