Benzyltrimethylammonium tetrachloroiodate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
B1543-5G
TCB1543-005GEA
234
USD
TCB1543-005G
Benzyltrimethylammonium tetrachloroiodate ≥95.0% (by titrimetric analysis)
Benzyltrimethylammonium tetrachloroiodate
[Chlorinating Reagent]
CAS Number: 121309-88-4
MDL Number: MFCD00191607
Molecular Formula: C10H16Cl4IN
Molecular Weight: 418.95
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 135
Formula:
C₁₀H₁₆Cl₄IN Melting Pt: 135 °C |
MDL Number:
MFCD00191607 CAS Number: 121309-88-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...