1,4-Bis(3-methoxyphenoxy)benzene ≥98.0%

Supplier: TCI America

Synonyms: Hydroquinone Bis(3-methoxyphenyl) Ether

B1486-5G
TCB1486-005GEA 221 USD
TCB1486-005G
1,4-Bis(3-methoxyphenoxy)benzene ≥98.0%
1,4-Bis(3-methoxyphenoxy)benzene

MDL Number: MFCD00142544
Molecular Formula: C20H18O4
Molecular Weight: 322.36
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 72


Other Product Names / Synonyms: Hydroquinone Bis(3-methoxyphenyl) Ether

Formula: C₂₀H₁₈O₄
MW: 322.36 g/mol
Melting Pt: 72 °C
MDL Number: MFCD00142544
CAS Number: 5024-84-0

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR