1,4-Bis(3-hydroxyphenoxy)benzene ≥96.0%
Supplier: TCI America
Synonyms:
Hydroquinone Bis(3-hydroxyphenyl) Ether
B1485-1G
TCB1485-001GEA
174
USD
TCB1485-001G
1,4-Bis(3-hydroxyphenoxy)benzene ≥96.0%
1,4-Bis(3-hydroxyphenoxy)benzene
CAS Number: 5085-95-0
MDL Number: MFCD00142543
Molecular Formula: C18H14O4
Molecular Weight: 294.31
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 126
Product Categories: Materials Chemistry, Monomers, Diphenyl Ethers (for High-Performance Polymer Research)
Other Product Names / Synonyms: Hydroquinone Bis(3-hydroxyphenyl) Ether
Formula:
C₁₈H₁₄O₄ MW: 294.31 g/mol Melting Pt: 126 °C |
MDL Number:
MFCD00142543 CAS Number: 5085-95-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...