1,3-Bis(2-isocyanato-2-propyl)benzene ≥97.0%

Supplier: TCI America

Synonyms: 1,3-Bis(1-isocyanato-1-methylethyl)benzene

B1442-500G B1442-25G
TCB1442-500GEA 150.01 USD
TCB1442-500G TCB1442-025G
1,3-Bis(2-isocyanato-2-propyl)benzene ≥97.0%
1,3-Bis(2-isocyanato-2-propyl)benzene

CAS Number: 2778-42-9
MDL Number: MFCD00142493
Molecular Formula: C14H16N2O2
Molecular Weight: 244.29
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Melting point (°C): -10
Flash Point (°C): 153
Specific Gravity (20/20): 1.08
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 1,3-Bis(1-isocyanato-1-methylethyl)benzene

Formula: C₁₄H₁₆N₂O₂
MW: 244.29 g/mol
Boiling Pt: 106 °C
Melting Pt: –10 °C
Density: 1.08 (20 °C)
Flash Pt: 153 °C
MDL Number: MFCD00142493
CAS Number: 2778-42-9
https://us.vwr.com/stibo/web/std.lang.all/32/34/28933234.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR