Glycidyl 4-tert-butylbenzoate ≥90.0%

Supplier: TCI America

Synonyms: 4-tert-Butylbenzoic Acid Glycidyl Ester

B1387-25G
TCB1387-025GEA 204 USD
TCB1387-025G
Glycidyl 4-tert-butylbenzoate ≥90.0%
Glycidyl 4-tert-butylbenzoate

CAS Number: 59313-58-5
MDL Number: MFCD00145184
Molecular Formula: C14H18O3
Molecular Weight: 234.30
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Flash Point (°C): 173
Specific Gravity (20/20): 1.08


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4-tert-Butylbenzoic Acid Glycidyl Ester

Formula: C₁₄H₁₈O₃
MW: 234.30 g/mol
Density: 1.08 (20 °C)
Flash Pt: 173 °C
MDL Number: MFCD00145184
CAS Number: 59313-58-5
https://us.vwr.com/stibo/web/std.lang.all/31/92/28933192.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR