5-tert-Butylisophthalic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America

B1380-25G
TCB1380-025GEA 322.68 USD
TCB1380-025G
5-tert-Butylisophthalic acid ≥99.0% (by GC, titration analysis)
5-tert-Butylisophthalic acid

CAS Number: 2359-09-3
MDL Number: MFCD00067047
Molecular Formula: C12H14O4
Molecular Weight: 222.24
Purity/Analysis Method: 99.0% (GC,T)
Form: Crystal

Formula: C₁₂H₁₄O₄
MDL Number: MFCD00067047
CAS Number: 2359-09-3
https://us.vwr.com/stibo/web/std.lang.all/31/87/28933187.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR