4-Amino-2-methylquinoline ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Aminoquinaldine2-Methylquinolin-4-amine

A0415-5G
TCA0415-5GEA 141.09 USD
TCA0415-5G
4-Amino-2-methylquinoline ≥97.0% (by GC, titration analysis)
4-Amino-2-methylquinoline

CAS Number: 6628-04-2
MDL Number: MFCD00006759
Molecular Formula: C10H10N2
Molecular Weight: 158.20
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Melting point (°C): 168


Other Product Names / Synonyms: 4-Aminoquinaldine

Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00006759
CAS Number: 6628-04-2
EINECS: 229-604-4
https://us.vwr.com/stibo/web/std.lang.all/78/53/28927853.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR