TCIAB1375

B1375-1G B1375-5G
TCB1375-001GEA 143.65 USD
TCB1375-001G TCB1375-005G
TCIAB1375
CAS Number: 25862-14-0
MDL Number: MFCD00059043
Molecular Formula: C14H14O3Se
Molecular Weight: 309.22
Purity/Analysis Method: 93.0% (HPLC)
Form: Crystal
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR