Bis(2-ethoxyethyl)amine ≥98.0%

Supplier: TCI America

Synonyms: 2,2'-Diethoxydiethylamine

B1364-25ML B1364-500ML
TCB1364-025MLEA 367.47 USD
TCB1364-025ML TCB1364-500ML
Bis(2-ethoxyethyl)amine ≥98.0%
Bis(2-ethoxyethyl)amine

CAS Number: 124-21-0
MDL Number: MFCD00039904
Molecular Formula: C8H19NO2
Molecular Weight: 161.25
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 200
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: 2,2'-Diethoxydiethylamine

Formula: C₈H₁₉NO₂
MW: 161.25 g/mol
Boiling Pt: 200 °C
Density: 0.89 (20 °C)
MDL Number: MFCD00039904
CAS Number: 124-21-0
https://us.vwr.com/stibo/web/std.lang.all/31/73/28933173.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR