Bis(dithiobenzil)nickel(II) ≥95.0% (by titrimetric analysis)
Supplier: TCI America
B1350-1G
TCB1350-001GEA
58.84
USD
TCB1350-001G
Bis(dithiobenzil)nickel(II) ≥95.0% (by titrimetric analysis)
Bis(dithiobenzil)nickel(II)
CAS Number: 28984-20-5
MDL Number: MFCD00059034
Molecular Formula: C28H20NiS4
Molecular Weight: 543.40
Purity/Analysis Method:
Form: Crystal
Lambda max.: 856 nm (Toluene)
Formula:
C₂₈H₂₀NiS₄ |
MDL Number:
MFCD00059034 CAS Number: 28984-20-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...