DL-2-Aminopimelic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: DL-2-Aminoheptanedioic acid

A0404-1G
TCA0404-001GEA 140.01 USD
TCA0404-001G
DL-2-Aminopimelic acid ≥98.0% (by titrimetric analysis)
DL-2-Aminopimelic acid

CAS Number: 627-76-9
MDL Number: MFCD00053314
Molecular Formula: C7H13NO4
Molecular Weight: 175.18
Purity/Analysis Method: 98.0% (T)
Form: Crystal

Formula: C₇H₁₃NO₄
MW: 175.18 g/mol
MDL Number: MFCD00053314
CAS Number: 627-76-9
https://us.vwr.com/stibo/web/std.lang.all/78/39/28927839.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR