4-Bromo-4'-pentylbiphenyl ≥99.0%

Supplier: TCI America

Synonyms: 4-Bromo-4'-pentyl-1,1'-biphenyl

B1305-5G
TCB1305-005GEA 44.64 USD
TCB1305-005G
4-Bromo-4'-pentylbiphenyl ≥99.0%
4-Bromo-4'-pentylbiphenyl

CAS Number: 63619-59-0
MDL Number: MFCD00060106
Molecular Formula: C17H19Br
Molecular Weight: 303.24
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Melting point (°C): 98


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Amyl-4'-bromobiphenyl

Formula: C₁₇H₁₉Br
MW: 303.2400 g/mol
Melting Pt: 98 °C
MDL Number: MFCD00060106
CAS Number: 63619-59-0
https://us.vwr.com/stibo/web/std.lang.all/31/33/28933133.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR