4-Bromo-4'-heptylbiphenyl ≥97.0%

Supplier: TCI America

Synonyms: 4-Bromo-4'-heptyl-1,1'-biphenyl

B1304-5G
TCB1304-005GEA 297 USD
TCB1304-005G
4-Bromo-4'-heptylbiphenyl ≥97.0%
4-Bromo-4'-heptylbiphenyl

CAS Number: 58573-93-6
MDL Number: MFCD00060107
Molecular Formula: C19H23Br
Molecular Weight: 331.30
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 95

Formula: C₁₉H₂₃Br
MW: 331.2899 g/mol
Melting Pt: 95 °C
MDL Number: MFCD00060107
CAS Number: 58573-93-6
https://us.vwr.com/stibo/web/std.lang.all/31/32/28933132.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR