8-Aminonaphthalene-1,3,6-trisulfonic acid disodium salt ≥99.0% (by HPLC, titration analysis)

Synonyms: Amino H Acid Disodium Salt8-Amino-1,3,6-naphthalenetrisulfonic Acid Disodium SaltKoch Acid Disodium Salt1-Naphthylamine-3,6,8-trisulfonic Acid Disodium Salt

A0398-25G
TCA0398-025GEA 95.54 USD
TCA0398-025G
8-Aminonaphthalene-1,3,6-trisulfonic acid disodium salt ≥99.0% (by HPLC, titration analysis)
8-Aminonaphthalene-1,3,6-trisulfonic acid disodium salt

Product Categories: Analytical Science, Fluorescent Dyes
Form: Crystal

Formula: C₁₀H₉NO₉S₃
MW: 427.32 g/mol
MDL Number: MFCD00070572
CAS Number: 5398-34-5

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR