4-Bromo-2,6-diethylaniline ≥98.0%
Supplier: TCI America
B1284-10G
TCB1284-010GEA
117
USD
TCB1284-010G
4-Bromo-2,6-diethylaniline ≥98.0%
4-Bromo-2,6-diethylaniline
CAS Number: 56746-19-1
MDL Number: MFCD00059266
Molecular Formula: C10H14BrN
Molecular Weight: 228.13
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 134
Specific Gravity (20/20): 1.33
Other Product Names / Synonyms: 2,6-Diethyl-4-bromoaniline
Formula:
C₁₀H₁₄BrN Boiling Pt: 134 °C Density: 1.33 (20 °C) |
MDL Number:
MFCD00059266 CAS Number: 56746-19-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...