Bis(trimethylenedithio)tetrathiafulvalene ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: Bis(1,3-propylenedithio)tetrathiafulvaleneBPDT-TTF

B1244-100MG
TCB1244-100MGEA 155.25 USD
TCB1244-100MG
Bis(trimethylenedithio)tetrathiafulvalene ≥97.0% (by HPLC)
Bis(trimethylenedithio)tetrathiafulvalene

CAS Number: 66946-49-4
MDL Number: MFCD00060080
Molecular Formula: C12H12S8
Molecular Weight: 412.71
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal
Melting point (°C): 250


Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: Bis(1,3-propylenedithio)tetrathiafulvalene, BPDT-TTF

Formula: C₁₂H₁₂S₈
MW: 412.71 g/mol
Melting Pt: 250 °C
MDL Number: MFCD00060080
CAS Number: 66946-49-4
https://us.vwr.com/stibo/web/std.lang.all/30/77/28933077.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR