Methyl docosanoate ≥90.0%

Supplier: TCI America

Synonyms: Behenic acid methyl ester Docosanoic acid methyl ester Methyl behenate

B1241-25G
TCB1241-025GEA 83.14 USD
TCB1241-025G
Methyl docosanoate ≥90.0%
Methyl docosanoate

CAS Number: 929-77-1
MDL Number: MFCD00009347
Molecular Formula: C23H46O2
Molecular Weight: 354.62
Purity/Analysis Method: 90.0% (GC)
Form: Crystal
Boiling point (°C): 393
Melting point (°C): 53
Flash Point (°C): 110


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Behenic Acid Methyl Ester, Docosanoic Acid Methyl Ester, Methyl Docosanoate

Formula: C₂₃H₄₆O₂
MW: 354.62 g/mol
Boiling Pt: 393 °C
Melting Pt: 53 °C
Flash Pt: 110 °C
MDL Number: MFCD00009347
CAS Number: 929-77-1
EINECS: 213-207-8
https://us.vwr.com/stibo/web/std.lang.all/30/74/28933074.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR