Benzenesulfonyl chloride ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: Benzene sulfochlorideBenzene sulphochloride

B1211-25G
TCB1211-025GEA 34.51 USD
TCB1211-025G
Benzenesulfonyl chloride ≥99.0% (by GC, titration analysis)
Benzenesulphonyl chloride

[for Determination of Hippuric Acid]
CAS Number: 98-09-9
MDL Number: MFCD00007426
Molecular Formula: C6H5ClO2S
Molecular Weight: 176.61
Purity/Analysis Method: 99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 252
Flash Point (°C): 135
Freezing point (°C): 15
Specific Gravity (20/20): 1.38

Formula: C₆H₅ClO₂S
MW: 176.62 g/mol
Boiling Pt: 252 °C
Melting Pt: 15 °C
Density: 1.38 (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00007426
CAS Number: 98-09-9
EINECS: 202-636-6
UN: 2225
ADR: 8,III
Merck Index: 13,01072
https://us.vwr.com/stibo/web/std.lang.all/30/47/28933047.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR