1-Benzyl-1-phenylhydrazine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1157-10G
TCB1157-010GEA 137.98 USD
TCB1157-010G
1-Benzyl-1-phenylhydrazine ≥98.0% (by titrimetric analysis)
1-Benzyl-1-phenylhydrazine

CAS Number: 614-31-3
MDL Number: MFCD00059186
Molecular Formula: C13H14N2
Molecular Weight: 198.27
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 218
Specific Gravity (20/20): 1.10

Formula: C₁₃H₁₄N₂
Boiling Pt: 218 °C
Density: 1.10 (20 °C)
MDL Number: MFCD00059186
CAS Number: 614-31-3
https://us.vwr.com/stibo/web/std.lang.all/30/00/28933000.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR