3-(4-tert-Butylphenyl)isobutyraldehyde ≥96.0%

Supplier: TCI America

Synonyms: 4-tert-Butyl-alpha-methylhydrocinnamaldehyde3-(4-(tert-Butyl)phenyl)-2-methylpropanal3-(4-tert-Butylphenyl)-2-methylpropionaldehyde

B1145-100ML B1145-25ML B1145-500ML
TCB1145-100MLEA 74.1 USD
TCB1145-100ML TCB1145-500ML TCB1145-025ML
3-(4-tert-Butylphenyl)isobutyraldehyde ≥96.0%
3-(4-tert-Butylphenyl)isobutyraldehyde

CAS Number: 80-54-6
MDL Number: MFCD00047655
Molecular Formula: C12H20O
Molecular Weight: 204.31
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 100
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: 4-tert-Butyl-alpha-methylhydrocinnamaldehyde, 3-(4-tert-Butylphenyl)-2-methylpropionaldehyde

Formula: C₁₂H₂₀O
MW: 204.31 g/mol
Boiling Pt: 150 °C
Density: 0.95 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00047655
CAS Number: 80-54-6
https://us.vwr.com/stibo/web/std.lang.all/29/89/28932989.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR