6-Amino-4-hydroxy-2-naphthalenesulfonic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-Amino-8-naphthol-6-sulphonic acid
, 7-Amino-1-naphthol-3-sulfonic acid
, 7-Amino-1-naphthol-3-sulphonic acid
, gamma acid
, 2-Amino-8-naphthol-6-sulfonic acid
CAS Number: 90-51-7
MDL Number: MFCD00003973
Molecular Formula: C10H9NO4S
Molecular Weight: 239.25
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: gamma Acid, 2-Amino-8-naphthol-6-sulfonic Acid, 7-Amino-1-naphthol-3-sulfonic Acid
Formula:
C₁₀H₉NO₄S MW: 239.25 g/mol |
MDL Number:
MFCD00003973 CAS Number: 90-51-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...