Acetoxime benzoate ≥98.0%

Synonyms: O-BenzoylacetoximeO-Benzoyloxime 2-PropanonePropan-2-one O-benzoyl oxime

A0066-25G
TCA0066-025GEA 147.9 USD
TCA0066-025G
Acetoxime benzoate ≥98.0%
Acetoxime benzoate

CAS Number: 942-89-2
MDL Number: MFCD00191697
Molecular Formula: C10H11NO2
Molecular Weight: 177.20
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 42


Other Product Names / Synonyms: O-Benzoylacetoxime, O-Benzoyloxime 2-Propanone

Formula: C₁₀H₁₁NO₂
MW: 177.20 g/mol
Boiling Pt: 120 °C
Melting Pt: 42 °C
MDL Number: MFCD00191697
CAS Number: 942-89-2
https://us.vwr.com/stibo/web/std.lang.all/75/08/28927508.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR