Bromoxylenol blue
Supplier: TCI America
B0998-25G
B0998-5G
TCB0998-25GEA
184.47
USD
TCB0998-25G
TCB0998-005G
Bromoxylenol blue
Bromoxylenol Blue
CAS Number: 40070-59-5
MDL Number: MFCD00005873
Molecular Formula: C23H20Br2O5S
Molecular Weight: 568.28
Form: Crystal
Color: Very Pale Yellow Red
Melting point (°C): 218
Lambda max.: 614 nm (H2O)
Other Product Names / Synonyms: 3',3''-Dibromo-p-xylenolsulfonephthalein
Formula:
C₂₃H₂₀Br₂O₅S Melting Pt: 218 °C |
MDL Number:
MFCD00005873 CAS Number: 40070-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...