Bis(2-hydroxy-5-chlorophenyl)sulfide ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,2'-Dihydroxy-5,5'-dichlorodiphenyl Sulfide
CAS Number: 97-24-5
MDL Number: MFCD00031479
Molecular Formula: C12H8Cl2O2S
Molecular Weight: 287.15
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 175
Product Categories: Materials Chemistry, Diphenyl Sulfides (for High-Performance Polymer Research)
Other Product Names / Synonyms: 2,2'-Dihydroxy-5,5'-dichlorodiphenyl Sulfide, Bis(5-chloro-2-hydroxyphenyl) Sulfide, 2,2'-Thiobis(4-chlorophenol)
Formula:
C₁₂H₈Cl₂O₂S MW: 287.15 g/mol Melting Pt: 175 °C |
MDL Number:
MFCD00031479 CAS Number: 97-24-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...