(Bromomethyl)chlorodimethylsilane ≥95.0%

Supplier: TCI America

Synonyms: BromomethyldimethylchlorosilaneBMDMCS

B0847-25G
TCB0847-025GEA 566.61 USD
TCB0847-025G
(Bromomethyl)chlorodimethylsilane ≥95.0%
(Bromomethyl)chlorodimethylsilane

CAS Number: 16532-02-8
MDL Number: MFCD00000503
Molecular Formula: C3H8BrClSi
Molecular Weight: 187.54
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 41
Specific Gravity (20/20): 1.39


Product Categories: Materials Chemistry, Silane Coupling Agents, Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: Bromomethyldimethylchlorosilane, BMDMCS

Formula: C₃H₈BrClSi
MW: 187.54 g/mol
Boiling Pt: 135 °C
Density: 1.39 (20 °C)
Flash Pt: 41 °C
MDL Number: MFCD00000503
CAS Number: 16532-02-8
https://us.vwr.com/stibo/web/std.lang.all/27/50/28932750.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR