Benzoyl cyanide ≥98.0%

Supplier: TCI America

Synonyms: Phenylglyoxylonitrile

B0835-25G
TCB0835-025GEA 69.02 USD
TCB0835-025G
Benzoyl cyanide ≥98.0%
Benzoyl cyanide

CAS Number: 613-90-1
MDL Number: MFCD00001837
Molecular Formula: C8H5NO
Molecular Weight: 131.13
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Flash Point (°C): 84
Freezing point (°C): 25


Other Product Names / Synonyms: Phenylglyoxylonitrile

https://us.vwr.com/stibo/web/std.lang.all/27/42/28932742.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR