Ethyl bis(2,4-dinitrophenyl)acetate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Bis(2,4-dinitrophenyl)acetic Acid Ethyl Ester

B0817-5G
TCB0817-005GEA 114.66 USD
TCB0817-005G
Ethyl bis(2,4-dinitrophenyl)acetate ≥98.0% (by HPLC)
Ethyl bis(2,4-dinitrophenyl)acetate

CAS Number: 5833-18-1
MDL Number: MFCD00051351
Molecular Formula: C16H12N4O10
Molecular Weight: 420.29
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 156


Product Categories: Analytical Science, Indicators (pH)
Other Product Names / Synonyms: Bis(2,4-dinitrophenyl)acetic Acid Ethyl Ester

Formula: C₁₆H₁₂N₄O₁₀
MW: 420.29 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00051351
CAS Number: 5833-18-1
https://us.vwr.com/stibo/web/std.lang.all/27/27/28932727.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR