8-Amino-2-naphthalenesulfonic acid ≥97.0% (by HPLC, titration analysis)

Synonyms: 1,7-Cleve's acid1-Naphthylamine-7-sulphonic acid1-Naphthylamine-7-sulfonic acid

A0356-500G A0356-25G
TCA0356-500GEA 510.6 USD
TCA0356-500G TCA0356-025G
8-Amino-2-naphthalenesulfonic acid ≥97.0% (by HPLC, titration analysis)
8-Amino-2-naphthalenesulphonic acid

CAS Number: 119-28-8
MDL Number: MFCD00004032
Molecular Formula: C10H9NO3S
Molecular Weight: 223.25
Purity/Analysis Method: 97.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 1,7-Cleve's Acid, 1-Naphthylamine-7-sulfonic Acid

Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
MDL Number: MFCD00004032
CAS Number: 119-28-8
EINECS: 204-311-4
https://us.vwr.com/stibo/web/std.lang.all/77/96/28927796.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR