TCIAB0781

B0781-25G
TCB0781-025GEA 80.44 USD
TCB0781-025G
TCIAB0781
CAS Number: 992-59-6
MDL Number: MFCD00012410
Molecular Formula: C34H28N6O6S2
Molecular Weight: 724.72
Form: Crystal
Color: Reddish Yellow

Other Product Names / Synonyms: Direct Red 2
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR