Crotyl alcohol ≥95.0%
Supplier: TCI America
Synonyms:
1-Hydroxy-2-butene
, 2-Buten-1-ol
B0696-25ML
TCB0696-025MLEA
126.89
USD
TCB0696-025ML
Crotyl alcohol ≥95.0%
Crotyl alcohol
CAS Number: 6117-91-5
MDL Number: MFCD00002923
Molecular Formula: C4H8O
Molecular Weight: 72.11
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 121
Flash Point (°C): 34
Specific Gravity (20/20): 0.85
Other Product Names / Synonyms: 2-Buten-1-ol, 3-Methylallyl Alcohol
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 121 °C Melting Pt: < –30 °C Density: 0.85 (20 °C) Flash Pt: 34 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002923 CAS Number: 6117-91-5 EINECS: 228-086-7 UN: 1987 ADR: 3,III Merck Index: 13,02629 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...