Xanthurenic acid ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
4,8-Dihydroxyquinoline-2-carboxylic acid
X0054-1G
TCX0054-001GEA
133.93
USD
TCX0054-001G
Xanthurenic acid ≥95.0% (by HPLC, titration analysis)
Xanthurenic acid
CAS Number: 59-00-7
MDL Number: MFCD00006754
Molecular Formula: C10H7NO4
Molecular Weight: 205.17
Purity/Analysis Method:
Form: Crystal
Color: Yellow
Other Product Names / Synonyms: 4,8-Dihydroxyquinoline-2-carboxylic Acid
Formula:
C₁₀H₇NO₄ MW: 205.17 g/mol Melting Pt: 297 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006754 CAS Number: 59-00-7 EINECS: 200-410-1 Merck Index: 12,10202 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...