Undecylbenzene ≥98.0%
Supplier: TCI America
Synonyms:
1-Phenylundecane
U0010-25ML
U0010-5ML
U0010-10ML
TCU0010-010MLEA
95.46
USD
TCU0010-010ML
TCU0010-025ML
TCU0010-5ML
Undecylbenzene ≥98.0%
Undecylbenzene
CAS Number: 6742-54-7
MDL Number: MFCD00026549
Molecular Formula: C17H28
Molecular Weight: 232.41
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 160
Melting point (°C): -5
Flash Point (°C): 110
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: 1-Phenylundecane
Formula:
C₁₇H₂₈ MW: 232.41 g/mol Boiling Pt: 160 °C Melting Pt: –5 °C Density: 0.86 (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026549 CAS Number: 6742-54-7 EINECS: 229-806-2 UN: 3082 ADR: 9,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...