2,3,5-Trimethylanisole ≥97.0%
Supplier: TCI America
Synonyms:
1-Methoxy-2,3,5-trimethylbenzene
T2456-5G
TCT2456-5GEA
60.88
USD
TCT2456-5G
2,3,5-Trimethylanisole ≥97.0%
2,3,5-Trimethylanisole
CAS Number: 20469-61-8
MDL Number: MFCD00142783
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method:
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 216
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: 1-Methoxy-2,3,5-trimethylbenzene
Formula:
C₁₀H₁₄O MW: 150.22 g/mol Boiling Pt: 216 °C Density: 0.97 (20 °C) |
MDL Number:
MFCD00142783 CAS Number: 20469-61-8 EINECS: 243-843-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...