2,3,5,6-Tetrafluorobenzaldehyde ≥94.0%

Supplier: TCI America
Warning

T1898-1G T1898-5G
TCT1898-5GEA 298.62 USD
TCT1898-5G TCT1898-1G
2,3,5,6-Tetrafluorobenzaldehyde ≥94.0%
2,3,5,6-Tetrafluorobenzaldehyde

CAS Number: 19842-76-3
MDL Number: MFCD00167387
Molecular Formula: C7H2F4O
Molecular Weight: 178.09
Purity/Analysis Method: 94.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 60
Flash Point (°C): 74
Specific Gravity (20/20): 1.55

Formula: C₇H₂F₄O
MW: 178.09 g/mol
Boiling Pt: 60 °C
Density: 1.55 (20 °C)
Flash Pt: 74 °C
MDL Number: MFCD00167387
CAS Number: 19842-76-3
https://us.vwr.com/stibo/web/std.lang.all/58/59/28995859.jpg

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR