2,3,4,5-Tetrafluoro-6-nitrobenzoic acid ≥98.0% (by titrimetric analysis)

Warning

T1862-25G T1862-5G
TCT1862-025GEA 323.42 USD
TCT1862-025G TCT1862-005G
2,3,4,5-Tetrafluoro-6-nitrobenzoic acid ≥98.0% (by titrimetric analysis)
2,3,4,5-Tetrafluoro-6-nitrobenzoic acid

CAS Number: 16583-08-7
MDL Number: MFCD02093916
Molecular Formula: C7HF4NO4
Molecular Weight: 239.08
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 139

Formula: C₇HF₄NO₄
MW: 239.08 g/mol
Melting Pt: 139 °C
MDL Number: MFCD02093916
CAS Number: 16583-08-7
https://us.vwr.com/stibo/web/std.lang.all/58/29/28995829.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR