1,3,5-Trimethylhexahydro-1,3,5-triazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Hexahydro-1,3,5-trimethyl-1,3,5-triazine

T1422-500ML T1422-25ML
TCT1422-500MLEA 500 USD
TCT1422-500ML TCT1422-025ML
1,3,5-Trimethylhexahydro-1,3,5-triazine ≥98.0% (by GC, titration analysis)
1,3,5-Trimethylhexahydro-1,3,5-triazine

CAS Number: 108-74-7
MDL Number: MFCD00006057
Molecular Formula: C6H15N3
Molecular Weight: 129.21
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 65
Flash Point (°C): 49
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: Hexahydro-1,3,5-trimethyl-1,3,5-triazine

Formula: C₆H₁₅N₃
MW: 129.21 g/mol
Boiling Pt: 65 °C
Density: 0.92 (20 °C)
Flash Pt: 49 °C
MDL Number: MFCD00006057
CAS Number: 108-74-7
EINECS: 203-612-8
https://us.vwr.com/stibo/web/std.lang.all/54/42/28995442.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR