trans-3,3,5-Trimethylcyclohexanol ≥95.0%
Supplier: TCI America
T0938-25G
TCT0938-025GEA
233.38
USD
TCT0938-025G
trans-3,3,5-Trimethylcyclohexanol ≥95.0%
trans-3,3,5-Trimethylcyclohexanol
CAS Number: 767-54-4
MDL Number: MFCD00066315
Molecular Formula: C9H18O
Molecular Weight: 142.24
Purity/Analysis Method:
Form: Crystal
Color: White
Boiling point (°C): 76
Melting point (°C): 58
Formula:
C₉H₁₈O MW: 142.24 g/mol Boiling Pt: 76 °C Melting Pt: 58 °C |
MDL Number:
MFCD00066315 CAS Number: 767-54-4 EINECS: 212-183-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...