cis-3,3,5-Trimethylcyclohexanol ≥96.0%

Supplier: TCI America

T0874-25G
TCT0874-025GEA 46.69 USD
TCT0874-025G
cis-3,3,5-Trimethylcyclohexanol ≥96.0%
cis-3,3,5-Trimethylcyclohexanol

CAS Number: 933-48-2
MDL Number: MFCD00070479
Molecular Formula: C9H18O
Molecular Weight: 142.24
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Color: White
Flash Point (°C): 88
Freezing point (°C): 35

Formula: C₉H₁₈O
MW: 142.24 g/mol
Boiling Pt: 196.5 °C (1013 hPa)
Melting Pt: 53…54 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00070479
CAS Number: 933-48-2
EINECS: 213-268-0
https://us.vwr.com/stibo/web/std.lang.all/49/79/28994979.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR