1,1,3-Trimethylcyclohexane ≥99.0%

Supplier: TCI America

T0827-5ML
TCT0827-005MLEA 278.03 USD
TCT0827-005ML
1,1,3-Trimethylcyclohexane ≥99.0%
1,1,3-Trimethylcyclohexane

CAS Number: 3073-66-3
MDL Number: MFCD00039462
Molecular Formula: C9H18
Molecular Weight: 126.24
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 138
Flash Point (°C): 18
Specific Gravity (20/20): 0.78

Formula: C₉H₁₈
MW: 126.24 g/mol
Boiling Pt: 138 °C
Melting Pt: –65.81 °C
Density: 0.78 (20 °C)
Flash Pt: 18 °C
MDL Number: MFCD00039462
CAS Number: 3073-66-3
EINECS: 221-347-6

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR