Tridecanenitrile ≥96.0%

Supplier: TCI America

Synonyms: Dodecyl cyanideLauryl cyanide

T0819-25G
TCT0819-025GEA 44.64 USD
TCT0819-025G
Tridecanenitrile ≥96.0%
Tridecanenitrile

CAS Number: 629-60-7
MDL Number: MFCD00039492
Molecular Formula: C13H25N
Molecular Weight: 195.35
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 152
Freezing point (°C): 9
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Dodecyl Cyanide, Lauryl Cyanide

Formula: C₁₃H₂₅N
Boiling Pt: 152 °C
Density: 0.83 (20 °C)
MDL Number: MFCD00039492
CAS Number: 629-60-7
EINECS: 211-097-6
https://us.vwr.com/stibo/web/std.lang.all/49/35/28994935.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR