7-Tetradecyne ≥97.0%

Supplier: TCI America

T0762-25ML T0762-5ML
TCT0762-005MLEA 182.62 USD
TCT0762-005ML TCT0762-25ML
7-Tetradecyne ≥97.0%
7-Tetradecyne

CAS Number: 35216-11-6
MDL Number: MFCD00041662
Molecular Formula: C14H26
Molecular Weight: 194.36
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 144
Specific Gravity (20/20): 0.79

Formula: C₁₄H₂₆
MW: 194.36 g/mol
Boiling Pt: 144 °C
Density: 0.79 (20 °C)
MDL Number: MFCD00041662
CAS Number: 35216-11-6
https://us.vwr.com/stibo/web/std.lang.all/48/94/28994894.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR