7-Tetradecene ≥97.0%

Supplier: TCI America

T0745-5ML
TCT0745-005MLEA 106.07 USD
TCT0745-005ML
7-Tetradecene ≥97.0%
7-Tetradecene

CAS Number: 10374-74-0
MDL Number: MFCD00009550
Molecular Formula: C14H28
Molecular Weight: 196.38
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 99
Specific Gravity (20/20): 0.77

Formula: C₁₄H₂₈
MW: 196.38 g/mol
Boiling Pt: 126…133 °C (0.05 mmHg)
Density: 0.77 (20 °C)
Flash Pt: 99 °C
MDL Number: MFCD00009550
CAS Number: 10374-74-0
EINECS: 233-815-7

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR