1,1,2,3-Tetrachloropropane ≥97.0%

Supplier: TCI America

T0707-10ML
TCT0707-010MLEA 453.51 USD
TCT0707-010ML
1,1,2,3-Tetrachloropropane ≥97.0%
1,1,2,3-Tetrachloropropane

CAS Number: 18495-30-2
MDL Number: MFCD00039343
Molecular Formula: C3H4Cl4
Molecular Weight: 181.87
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 179
Specific Gravity (20/20): 1.53

Formula: Cl₂HCCHClCH₂Cl
MW: 181.88 g/mol
Boiling Pt: 179 °C
Density: 1.53 (20 °C)
MDL Number: MFCD00039343
CAS Number: 18495-30-2
EINECS: 242-379-7

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR