Trimethylthiourea ≥98.0% (by total nitrogen basis)

Supplier: TCI America

T0667-100G T0667-500G
TCT0667-500GEA 180 USD
TCT0667-500G TCT0667-100G
Trimethylthiourea ≥98.0% (by total nitrogen basis)
Trimethylthiourea

CAS Number: 2489-77-2
MDL Number: MFCD00059148
Molecular Formula: C4H10N2S
Molecular Weight: 118.20
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 84

Formula: C₄H₁₀N₂S
Melting Pt: 84 °C
MDL Number: MFCD00059148
CAS Number: 2489-77-2
EINECS: 219-644-0
https://us.vwr.com/stibo/web/std.lang.all/48/36/28994836.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR